Brady® i75 R4400 Ribbon for the i7500 Label Printer
Supplier: Brady Worldwide
176777 176778 176779 176780 176781 176782 176783 176784 176785 176786 176787
77775-072RL 0 USD
77775-072 77775-074 77775-076 77775-078 77775-080 77775-082 77775-084 77775-086 77775-088 77775-090 77775-092
Brady® i75 R4400 Ribbon for the i7500 Label Printer
Printer Ribbons
i75-Series ribbon with LabelSense™ Technology warns user if ribbon is incompatible with installed i75-Series label material. Automatically prints at pre-tested heat and speed, ensuring correct print quality on the first label with no setting adjustments or trial-and-error print testing.
  • Ribbon in cartridge format
  • Ribbin ink wind direction: ink side out
  • Polyester material
i75-series ribbon is self-centering with ink-side always correctly facing the label. One-color ribbon for printing a single color onto a label. R4400 Series high-density colored resin
More Product Information
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable