1-Nonadecanol 98%

Supplier: Ambeed
Warning

Synonyms: Nonadecyl alcohol

A366305-1G A366305-1MG A366305-250MG A366305-5G A366305-25G A366305-100MG
77240-188EA 18 USD
77240-188 77240-190 77240-192 77240-194 77616-600 77746-732
1-Nonadecanol 98%
1-Nonadecanol

Caution: for Research Use Only

Formula: H₃C(CH₂)₁₇CH₂OH
MDL Number: MFCD00002824
CAS Number: 1454-84-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR