QC Standard 19: As, Be, Ca, Cd, Co, Cr, Cu, Fe, Mg, Mn, Mo, Ni, Pb, Sb, Se, Ti, Tl, V, Zn @ 100 µg/mL in 5% HNO3, tr. F-, tr. Tartaric Acid, VHG, LGC Standards
Supplier: LGC Standards
VHG-QC19-100
77697-839EA 273.2 USD
77697-839
QC Standard 19: As, Be, Ca, Cd, Co, Cr, Cu, Fe, Mg, Mn, Mo, Ni, Pb, Sb, Se, Ti, Tl, V, Zn @ 100 µg/mL in 5% HNO3, tr. F-, tr. Tartaric Acid, VHG, LGC Standards
Standards Cationic Standards Multi-Element Standards