ICH/USP Parenteral Combined-2 Target Elements: Au @ 100; Ir, Os, Pd, Pt, Rh, Ru @ 10 µg/mL in 15% HCl, VHG, LGC Standards
Supplier: LGC Standards
VHG-ICH-USP-TELG-100
77697-541EA 247.65 USD
77697-541
ICH/USP Parenteral Combined-2 Target Elements: Au @ 100; Ir, Os, Pd, Pt, Rh, Ru @ 10 µg/mL in 15% HCl, VHG, LGC Standards
Standards Cationic Standards Multi-Element Standards