ICP-AES CRQL Solution 2: Ca, K, Mg, Na @ 5000 µg/mL in 5% HNO3, VHG, LGC Standards
Supplier: LGC Standards
VHG-CRQL2AES-100
77697-480EA 221.9 USD
77697-480
ICP-AES CRQL Solution 2: Ca, K, Mg, Na @ 5000 µg/mL in 5% HNO3, VHG, LGC Standards
Standards Cationic Standards Multi-Element Standards