Triamcinolone, LoGiCal, LGC Standards

Supplier: LGC Standards

LGCFOR0198.01
77693-685EA 215.25 USD
77693-685
Triamcinolone, LoGiCal, LGC Standards
Standards Organic Standards
New Product

Order Now

SPECIFICATIONS

Environmentally Preferable
CAS No. 124-94-7
Formula C₂₁H₂₇FO₆
Molecular Weight 394.44 g/mol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR