Chromium Standard: Cr @ 1000 µg/mL in 5% HNO3, VHG, LGC Standards
Supplier: LGC Standards
VHG-PCRN-100 VHG-PCRN-250 VHG-PCRN-500
77693-243EA 55.2 USD
77693-243 77693-244 77693-245
Chromium Standard: Cr @ 1000 µg/mL in 5% HNO3, VHG, LGC Standards
Standards Cationic Standards Chromium Standards
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable