Gold ≥99.999%, disk, as rolled, Diameter 29 mm

Supplier: GOODFELLOW CORPORATION

1000016094 1000046365 1000044265
77680-709EA 5180.02 USD
77680-709 77681-607 77681-466
Gold ≥99.999%, disk, as rolled, Diameter 29 mm
Gold
Formula: Au
Boiling Pt: 3080 °C
Melting Pt: 1064.4 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR