Gold ≥99.95%, disk, as rolled, Diameter 50.8 mm

Supplier: GOODFELLOW CORPORATION

1000031081 1000030890 1000031500 1000032001 1000030632 1000030755 1000031954 1000032111 1000044761
77680-950EA 3447.91 USD
77680-950 77680-910 77681-033 77681-162 77680-838 77680-880 77681-148 77681-191 77681-485
Gold ≥99.95%, disk, as rolled, Diameter 50.8 mm
Gold
Formula: Au
Boiling Pt: 3080 °C
Melting Pt: 1064.4 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR