Gold ≥99.9%, disk, Diameter 50 mm

Supplier: GOODFELLOW CORPORATION

1000030321 1000030423 1000030528 1000030377 1000003645 1000031834 1000030823 1000031643
77680-743EA 737.53 USD
77680-743 77680-784 77680-810 77680-769 77680-694 77681-107 77680-896 77681-062
Gold ≥99.9%, disk, Diameter 50 mm
Gold
Formula: Au
Boiling Pt: 3080 °C
Melting Pt: 1064.4 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR