Gold ≥99.95%, disk, as rolled, Diameter 8 mm

Supplier: GOODFELLOW CORPORATION

1000031091 1000003700 1000025022 1000031964 1000032007 1000030640 1000030763 1000031713 1000031295 1000032116
77680-954EA 491.43 USD
77680-954 77680-695 77680-725 77681-152 77681-166 77680-842 77680-884 77681-071 77681-004 77681-194
Gold ≥99.95%, disk, as rolled, Diameter 8 mm
Gold
Formula: Au
Boiling Pt: 3080 °C
Melting Pt: 1064.4 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR