Gold ≥99.99%, disk, as rolled, Diameter 40 mm

Supplier: GOODFELLOW CORPORATION

1000030666 1000031917 1000030698 1000030563 1000031574 1000032080
77680-851EA 886.38 USD
77680-851 77681-133 77680-864 77680-821 77681-045 77681-175
Gold ≥99.99%, disk, as rolled, Diameter 40 mm
Gold
Formula: Au
Boiling Pt: 3080 °C
Melting Pt: 1064.4 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR