BioReady Buffers

Supplier: NANOCOMPOSIX, INC.

BLBR-100M BLBR-1L BLBR-500M RUPR-100M RUPR-1L RUPR-500M RXKR-100M RXKR-1L RXKR-500M RXNR-100M RXNR-1L RXNR-500M RXPR-100M CDPR-100M CDPR-1L CDPR-500M DSKR-100M DSKR-1L DSKR-500M RXPR-1L RXPR-500M
77680-214EA 175.75 USD
77680-214 77680-216 77680-218 77680-502 77680-504 77680-506 77680-508 77680-510 77680-512 77680-514 77680-516 77680-518 77680-520 77680-228 77680-230 77680-232 77680-234 77680-236 77680-238 77680-522 77680-524
BioReady Buffers
Buffers Electrophoresis Buffers Running Buffers

The same high quality conjugation reaction buffers, block buffer, and conjugate diluent for covalent conjugation found in our high sensitivity gold conjugation kit, now available in catalog and bulk volumes.


  • Convenient one-stop shopping for conjugation buffers and nanoparticles
  • Save time on formulating buffers and writing SOPs
  • Ensure consistent quality and supply for your conjugation


BioReady Block Covalent Buffer
4 mM sodium tetraborate, 1% BSA, 0.05% sodium azide, pH 8.2
BioReady Running Buffer
1X PBS, 1% Tween 20, pH 7.4


BioReady Sodium Phosphate Reaction Buffer
5mM sodium phosphate, 0.5% PEG20K, pH 7.4


BioReady Conjugate Diluent
0.5X PBS, 0.5% BSA, 0.5% casein, 1% Tween 20, 0.05% sodium azide, pH 8.0


BioReady Potassium Phosphate Reaction Buffer
5 mM potassium phosphate, 0.5% PEG20K, pH 7.4


BioReady PBS Reaction Buffer
0.01X PBS, 0.5% PEG20K, pH 7.4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR