BioPure Silver Nanospheres 1 mg/ml Citrate (bare/displaceable surface), nanoComposix
Supplier: NANOCOMPOSIX, INC.
AGCB200-30M AGCB100-30M AGCB10-30M AGCB50-30M AGCB5-30M AGCB20-30M AGCB30-30M AGCB40-30M AGCB60-30M AGCB80-30M
103819-596EA 3740 USD
103819-596 77679-666 77679-674 77679-706 77679-714 77679-682 77679-690 77679-698 77679-722 77679-730
BioPure Silver Nanospheres 1 mg/ml Citrate (bare/displaceable surface), nanoComposix
Beads Microcarriers
Purified, concentrated, high quality silver nps; sterile and endotoxin-free. Bare citrate surface.
  • High quality nanoparticles, precisely engineered and highly characterized
  • Unagglomerated and monodisperse
  • High concentration materials (1 mg/ml, 9.27 mm, 1000 ppm, ~40 to 135 OD [depending on size])
  • Extensively purified to remove residual reactants
  • Manufactured sterile; provided in ultra to pure (Milli to Q) DI water
  • BioPure mateirals ideal for in to vitro and in to vivo experiments, nanotoxicology
  • "Bare" citrate surface is stable and displaceable
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable