6-Oxopiperidine-3-carboxylic acid 98

Supplier: Ambeed
Warning

A244771-5G
77672-188EA 218 USD
77672-188
6-Oxopiperidine-3-carboxylic acid 98
6-Oxopiperidine-3-carboxylic acid

Caution: for Research Use Only

Formula: C₆H₉NO₃
MW: 143.14 g/mol
MDL Number: MFCD08059985
CAS Number: 22540-50-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR