Triethyl citrate 99+%

Supplier: Ambeed
Warning

Synonyms: Triethyl 2-hydroxypropane-1,2,3-tricarboxylate, Hydroxytricarballylic acid triethyl ester, Citric acid triethyl ester

A485424-100G A485424-1000G A485424-25G
77602-098EA 13.33 USD
77602-098 77669-164 77669-166
Triethyl citrate 99+%
Triethyl citrate

Caution: for Research Use Only

Formula: C₁₂H₂₀O₇
MDL Number: MFCD00009201
CAS Number: 77-93-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR