Classic Crimped Wire Mesh Sterilization Baskets, Tilted Handles
Supplier: WORLD PRECISION INSTRUMENTS LLC
WP-4403ER WP-4404ER WP-4405ER WP-4406ER WP-4426ER WP-4427ER WP-4428ER WP-4429ER WP-4414ER WP-4415ER WP-4416ER WP-4417ER WP-4422ER WP-4423ER WP-4424ER WP-4425ER WP-4430ER WP-4431ER WP-4432ER WP-4433ER WP-4434ER WP-4435ER WP-4436ER WP-4437ER WP-4438ER WP-4401ER WP-4402ER WP-4411ER WP-4412ER WP-4413ER WP-4439ER WP-4440ER WP-4441ER WP-4442ER WP-4443ER WP-4407ER WP-4408ER WP-4409ER WP-4410ER WP-4418ER WP-4419ER WP-4420ER WP-4421ER WP-4444ER WP-4445ER
77665-876EA 79 USD
77665-876 77665-882 77665-886 77665-892 77665-996 77666-002 77666-006 77666-010 77665-938 77665-944 77665-948 77665-952 77665-976 77665-980 77665-986 77665-992 77666-016 77666-022 77666-026 77666-030 77666-034 77666-038 77666-044 77666-048 77666-054 77665-866 77665-872 77665-922 77665-928 77665-934 77666-058 77666-062 77666-066 77666-070 77666-074 77665-898 77665-902 77665-906 77665-914 77665-958 77665-962 77665-966 77665-970 77666-080 77666-086
Classic Crimped Wire Mesh Sterilization Baskets, Tilted Handles
Baskets Wire Baskets
Stainless steel classic crimped wire mesh sterilization basket with tilted handles.
  • Manufactured with stainless steel 304
  • Electropolished
  • Burr free to protect wraps and prevent injury
  • Smooth rounded corners
  • Tilt handles
  • Single frame, double frame and detention frame lids available
Benefits:
Meets all the requirements of automatic washing and sterilization for all types of instruments
Top and bottom frame diameter: 5.0 mm
Crimped mesh with mesh openings of 6 mm, wire diameter of 1.0 mm
Smooth corners and tilt handles
Applications:
Instrument cleaning,More Product Information