NanoFil™ Needles for Gas-Tight Injection System
Supplier: WORLD PRECISION INSTRUMENTS LLC
NF26BV-2
77664-904PK 101.65 USD
77664-904
NanoFil™ Needles for Gas-Tight Injection System
Syringe Needles
Needles for use with NanoFil™ gas-tight injection system.
  • Beveled tips that are shorter and more durable
  • Offers better penetration than standard removable needles
  • Made up of stainless steel
Beveling the needle on three sides creates an actual point on the needle that penetrates better and is more durable than a single beveled needle. In addition, the distance from the point to the upper opening of the tip is reduced allowing for less penetration into the tissue and therefore less tissue damage.
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable