Cell Culture Cup Chambers for TEER Measurement
Supplier: WORLD PRECISION INSTRUMENTS LLC
ENDOHM-12G ENDOHM-6G ENDOHM-24G-SNAP
77664-246EA 2250 USD
77664-246 77664-250 77664-248
Cell Culture Cup Chambers for TEER Measurement
Culture Dishes Cell Culture Dishes
These cell culture cup chambers used in TEER measurement for removable culture cup systems using EVOM2 meters for endothelial and epithelial cell cultures.
  • Adjustable apical electrode height
  • Crystal clear glass chamber allows visualization of apical electrode positioning
  • New insert holder with 120º tri-supports for three leg inserts
  • Compatible with EVOM, EVOM2™ and Millicell® ERS and ERS2
  • For EndOhms used with the new EVOM™ Manual (or EVOM3), see EVOM™ Electrodes for TEER
EndOhm chambers provide reproducible resistance measurements of endothelial andMore Product Information