Anesthesia Facemask for Cunningham Stereotaxic Frame Adapters
Supplier: WORLD PRECISION INSTRUMENTS LLC
502063-MGM 502063-SGM 502063-SGM
MSPP-502063MGMEA 628 USD
MSPP-502063MGM MSPP-502063SGM 77663-050
Anesthesia Facemask for Cunningham Stereotaxic Frame Adapters
Pumps Pump Drives Peristaltic Pump Drives
Our custom toothbar for Cunningham adaptor is designed to accommodate use of a soft gas mask. The included mask securely fits the toothbar to eliminate gas leakage.
  • Facemask custom fitted to a toothbar that fits a cunningham stereotaxic adapter
  • Anatomically correct design for a tight fit for mice and rats
  • Zero leakage
  • Zero dead space
  • Ports swivel 360°
  • Optically clear
  • Designed to provide access to eyes and head of the rodent
  • Mask provides stability of the animal for examinations and surgery
  • Includes OC-tubing and OC-adapter (requires MSPP-EZ-1130)