Multi-Barrel® Glass Capillaries
Supplier: WORLD PRECISION INSTRUMENTS LLC
2B150F-4 3B120F-4 3B120F-6 2B150F-6 7B100F-4 7B100F-6 7B120F-4 5B120F-4
77662-038PK 94 USD
77662-038 77662-090 77662-092 77662-040 77663-968 77663-970 77663-972 77663-906
Multi-Barrel® Glass Capillaries
Pipettes Microcapillaries
Multi-barrel configurations are designed especially for microiontophoresis. Because the capillaries are fused together during manufacture, you will not need to twist them while pulling to seal the tips together. An inner filament in each barrel makes filling easy and fast.
  • All glass capillaries barrels include a filament
  • O.D/I.D tolerance: ±0.1 mm
  • Length tolerance: ±1 mm
  • Recommended to use with glass handling forceps
  • Superior pricing
  • Most glass orders ship within 48 hours
  • Applications are microinjection, electrophysiology, patch clamp and fluid handling
More Product Information