Single-Barrel Standard Glass Capillaries
Supplier: WORLD PRECISION INSTRUMENTS LLC
1B200-4 1B200-6 1B200F-4 1B200F-6 1B100-3 1B100-4 1B100-6 1B100F-3 1B100F-4 1B100F-6 1B120-3 1B120-4 1B120-6 1B120F-3 1B120F-4 1B120F-6 1B150-3 1B150-4 1B150-6 1B150F-3 1B150F-4 1B150F-6
77662-016PK 92 USD
77662-016 77662-018 77662-020 77662-022 77661-980 77661-982 77661-984 77661-986 77661-988 77661-990 77661-992 77661-994 77661-996 77661-998 77662-000 77662-002 77662-004 77662-006 77662-008 77662-010 77662-012 77662-014
Single-Barrel Standard Glass Capillaries
Pipettes Microcapillaries
These standard glass capillaries are made with quality, borosilicate glass and used for applications such as: Microinjection, electrophysiology, patch clamp, and fluid handling.
  • Quality borosilicate glass capillaries
  • Large variety available
  • Some varieties are fire polished
  • OD/ID tolerance: ±0.1 mm
  • Length tolerance: ±1 mm
  • Recommended to use with glass handling forceps
Fire-Polished glass capillaries are easier to insert into microelectrode holders without damaging the gasket. More importantly, fire-polished glass won’t scratch the chloridized wire usedMore Product Information