Tyrell Hook
Supplier: WORLD PRECISION INSTRUMENTS LLC
14136-G
77661-800EA 60 USD
77661-800
Tyrell Hook
Dissection Sets
A Tyrell hook is a surgical instrument commonly used in neurosurgery, often in the removal of brain tumors or other types of abnormal tissue. Named after its inventor, Dr. Ivan Tyrell, who was a British neurosurgeon, the Tyrell hook is a long, slender instrument with a curved hook at the end.
  • Sharp, micro tip
  • Premium German steel
The hook allow the surgeon to gently manipulate and move tissue during the surgical procedure, while minimizing the risk of damage to surrounding structures such as blood vessels or nerves.
A Tyrell hook may be used toMore Product Information