Sartolab® P20 and P20 Plus Pressure Filtration Units
Supplier: Sartorius
18076----------N 18089----------D 18090----------D 18091----------D 18092----------D
77661-238PK 1010 USD
77661-238 77661-240 77661-242 77661-244 77661-246
Sartolab® P20 and P20 Plus Pressure Filtration Units
Filters Capsule Filters
Sartolab® P20 and P20 Plus are ready-to-use pressure filter units for the clarification of 100 ml to 10 L particle-high-loaded solutions.
  • High flow rates
  • Large surface area (20 cm²)
  • Protein-preserving, low-binding membrane
  • Optimized membrane for low dead volume
  • Versions available with a prefilter for high particle load solutions
Sartolab® P20 and P20 Plus pressure filter units are available with a 0.2 µm or 0.45 µm polyethersulfone membrane, protected by a prefilter made of high purity quartz microfibers. Different configurations are available:More Product Information