VWR® Mini Shakers, 230 V
Supplier: VWR International
77623-746 77623-750 77623-754 77623-758
77623-746EA 984 USD
77623-746 77623-750 77623-754 77623-758
VWR® Mini Shakers, 230 V
Shakers and Mixers Orbital Shakers
VWR® mini shakers feature variable-speed, microprocessor control, providing reliable, uniform mixing action. Ideal for cell cultures, solubility studies, extraction procedures, diagnostic tests, general mixing, hybridization, genetics research, bacterial suspensions, staining/destaining, and washing procedures.
  • 230 V models
  • Choice of models with either 3 or 20 mm orbit
  • LCD display for speed and time
  • Includes platform and mat; ready to attach a variety of optional accessories
  • Ideal for use in non-condensing incubators, CO₂ incubators, or cold rooms
  • RS232
More Product Information