VWR® 550 Benchtop pH Meters
Supplier: VWR International
77619-150 77619-152 77619-154 77619-156 77619-158 77619-160 77619-162 77619-164 77619-166
77619-150EA 377.54 USD
77619-150 77619-152 77619-154 77619-156 77619-158 77619-160 77619-162 77619-164 77619-166
VWR® 550 Benchtop pH Meters
Electrochemistry Meters pH and Multi-Parameter Meters
The VWR-PH550 meter provide accurate and reliable measurements of pH, mV and temperature in any environment.
  • High resolution LCD Display (6.0")
  • 1 to 3 point calibration
  • Automatic identification of three pH standard buffer solutions (4.01, 7.00, and 10.01 pH)
  • IP 54 protection
  • Automatic or manual temperature compensation capabilities
  • Auto-read and continuous-read measurement options
  • Ability to save 50 data sets
  • Displays electrode status (electrode slope)
  • Three-year meter warranty
The easy to use VWR-PH550 benchtop meters provide accurate measurementsMore Product Information