VWR® M700 Portable pH/Ion/Conductivity/Dissolved Oxygen Meters
Supplier: VWR International
77619-094 77619-096 77619-098 77619-100
77619-094EA 1041.11 USD
77619-094 77619-096 77619-098 77619-100
VWR® M700 Portable pH/Ion/Conductivity/Dissolved Oxygen Meters
Electrochemistry Meters pH and Multi-Parameter Meters
The VWR-M700 portable multi-parameter meter provides accurate and reliable measurements of pH, conductivity, ion, and dissolved oxygen within an easy to use meter.
  • High resolution LCD display (3.5")
  • Five-point calibration for pH with selectable pH buffer group
  • Built-in ion methods including F⁻, NH₄⁺ and NA⁺
  • IP 65 waterproof
  • Automatic or manual temperature compensation capabilities
  • Auto-read, timed-read, and continuous-read measurement options
  • Three-point conductivity calibration with Standard Recognition
  • Dissolved oxygen calibration supports air-saturated
More Product Information