VWR® 900 Portable Ion/pH Meters
Supplier: VWR International
77619-030 77619-032 77619-034 77619-036 77618-922 77619-050 77619-052 77619-054 77619-056 77619-058 77619-028 77619-038 77619-040 77619-042 77619-044 77619-046 77619-048
77619-030EA 1652.91 USD
77619-030 77619-032 77619-034 77619-036 77618-922 77619-050 77619-052 77619-054 77619-056 77619-058 77619-028 77619-038 77619-040 77619-042 77619-044 77619-046 77619-048
VWR® 900 Portable Ion/pH Meters
Electrochemistry Meters pH and Multi-Parameter Meters
The VWR-I900 meter provide accurate and reliable measurements of ISE, pH, mV and temperature in any environment.
  • High resolution color touchscreen (4.3")
  • 1 to 8 point calibration with standard recognition for pH
  • Built-in ion methods including F⁻, CL⁻, K⁺, and Na⁺
  • Selectable pH buffer groups including NIST, DIN, GB, USA
  • IP 65 waterproof
  • Ability to save 1000 sets of measurement results (GLP-compliant)
  • Rechargeable battery
  • Three-year meter warranty
Automatic or Manual temperature compensation capabilities. Auto-read, timed-read, and continuous-readMore Product Information