VWR® 750 Benchtop pH/Ion Meters
Supplier: VWR International
77618-920 77618-996 77618-998 77619-000 77619-002 77619-004 77619-006 77619-008 77619-010 77619-012 77619-014 77619-016 77619-018 77619-020 77619-022 77619-024 77619-026
77618-920EA 1547.13 USD
77618-920 77618-996 77618-998 77619-000 77619-002 77619-004 77619-006 77619-008 77619-010 77619-012 77619-014 77619-016 77619-018 77619-020 77619-022 77619-024 77619-026
VWR® 750 Benchtop pH/Ion Meters
Electrochemistry Meters pH and Multi-Parameter Meters
The VWR-I750 meter provides accurate and reliable measurements of pH, mV, ion conecentration, and pX.
  • High resolution LCD display (5.7")
  • One to five points pH and ion calibration with standard recognition
  • Selectable pH buffer groups including NIST, DIN, GB, USA
  • Built-in ion methods including F⁻, Br⁻, NO³⁻, and Ag⁺
  • Ability for user to define ion measurement method
  • IP54 rating
  • Automatic or manual temperature compensation capabilities
  • Auto-read, timed-read, and continuous-read measurement options
  • Data capacity of 500 sets (GLP-compliant)
  • Automatic electrode
More Product Information