VWR® 950 Benchtop Dissolved Oxygen Meters
Supplier: VWR International
77618-942 77618-944
77618-942EA 1502.65 USD
77618-942 77618-944
VWR® 950 Benchtop Dissolved Oxygen Meters
Electrochemistry Meters Oxygen Meters
The VWR-DO950 meter provide accurate and reliable measurements of dissolved oxygen concentration and dissolved oxygen saturation.
  • High resolution color touch display (7.0")
  • Air-saturated water or zero oxygen calibration capabilities
  • Automatic or manual temperature compensation capabilities
  • Auto-read, timed-read, and continuous-read measurement options
  • Data capacity of 1000 sets for each parameter (GLP-compliant)
  • Auto barometric pressure compensation
  • For use with DO1 Polarographic DO Electrode (77618-816)
The easy to use VWR-DO950 portable meters
More Product Information