VWR® Premium Series Upright Ultra-Low Temperature Freezers
Supplier: VWR International
77618-802 77618-804 77618-762 77618-764 77618-766 77618-768 77618-774 77618-776 77618-778 77618-780 77618-786 77618-788 77618-790 77618-792 77618-798 77618-800
77618-802EA 16858.38 USD
77618-802 77618-804 77618-762 77618-764 77618-766 77618-768 77618-774 77618-776 77618-778 77618-780 77618-786 77618-788 77618-790 77618-792 77618-798 77618-800
VWR® Premium Series Upright Ultra-Low Temperature Freezers
Freezers Ultra Low Temperature Freezers
The VWR® Premium Series ULT freezers provide the power of performance with energy efficiency for a greener tomorrow.
  • Operate between -50 to -86 °C
  • 4 storage capacities: (400, 500, 600, 700 2" cryoboxes)
  • ENERGY STAR® certified
  • Easy-to-use touch button interface
  • Vacuum insulation panels and high density water-blown foam
  • USB for easy export of recorded temperatures and events
  • 5 year warranty parts and labor
Experience the power of these premium ULT freezers, meticulously engineered to excel in the most demanding laboratory settings. With preciseMore Product Information