5-Aza-2'-deoxycytidine 99%

Supplier: Ambeed
Warning

Synonyms: 5-Aza-2′-deoxycytidine, Decitabine, 5-Aza-1-(2-deoxy-beta-D-ribofuranosyl)cytosine, 4-Amino-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1,3,5-triazin-2(1H)-one, 2′-Deoxy-5-azacytidine 4-Amino-1-(2-deoxy-β-D-ribofuranosyl)-1,3,5-triazin-2(1H)-one Decitabine, 4-Amino-1-(2-deoxy-beta-D-ribofuranosyl)-1,3,5-triazin-2(1H)-one

A156645-100MG A156645-10MG A156645-1MG A156645-1G A156645-250MG A156645-50MG A156645-5G
76997-554EA 25 USD
76997-554 76997-556 76997-558 77549-988 77600-238 77600-240 77671-210
5-Aza-2'-deoxycytidine 99%
5-Aza-2'-deoxycytidine

Caution: for Research Use Only

Formula: C₈H₁₂N₄O₄
MDL Number: MFCD00043011
CAS Number: 2353-33-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR