LabExact® Trupor® Nylon High Flow, Low Back Pressure Filter Membranes
Supplier: I.W. TREMONT CO., INC.
LETNY045090 LETNY045142 LETNY020025 LETNY020047 LETNY020090 LETNY020142 LETNY045025 LETNY045047
77591-606BX 385.92 USD
77591-606 77591-608 77591-594 77591-596 77591-598 77591-600 77591-602 77591-604
LabExact® Trupor® Nylon High Flow, Low Back Pressure Filter Membranes
Filters Membrane Filters
H&V Trupor® Nylon membranes utilize nanoscale technology in a composite design to achieve precisely regulated porosities with higher loading capacity and faster flow. No adhesives are utilized, which significantly reduces trace extractables.
  • No additives are utilized, which significantly reduces trace extractables
  • Trupor® membrane filters demonstrate back pressure reduction by >50% versus comparable porosity cast membranes
  • Trupor® Nylon membrane filters demonstrate loading capacity >40% versus comparable porosity cast membrane
  • Larger sample
More Product Information
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable