LabExact® Cellulose Acetate Syringe Filters
Supplier: I.W. TREMONT CO., INC.
LEES10081 LEES10082 LEES10083 LEES10084 LEES10611 LEES10612
77590-986PK 128.29 USD
77590-986 77590-988 77590-990 77590-992 77591-118 77591-120
LabExact® Cellulose Acetate Syringe Filters
Filters Syringe Filters
LabExact® disposable non-sterile Cellulose Acetate hydrophilic syringe filters have very low binding capacity for proteins and used for filtration and clarification of biological fluids.
  • Cellulose acetate is nitrate-free making it suitable for groundwater and EPA ASTM’s
  • Use for filtration under sterile conditions or filtration and clarification of biological fluids, sterilization of media additives and serum and sample preparation of aqueous or protein-based HPLC solutions
  • Use for applications requiring maximum sample recovery
  • Designed to sterilize and
More Product Information