LabExact® Grade B 1.0 µm Binderless Glass Microfiber Filters
Supplier: I.W. TREMONT CO., INC.
LEB2500 LEB3500 LEB3700 LEB4250 LEB4700 LEB5500 LEB7000 LEB9000 LEBS810 LEB1100 LEB1250 LEB1500 LEB2100 LEB2400
77590-466BX 30.75 USD
77590-466 77590-468 77590-470 77590-472 77590-474 77590-476 77590-478 77590-480 77590-486 77590-456 77590-458 77590-460 77590-462 77590-464
LabExact® Grade B 1.0 µm Binderless Glass Microfiber Filters
Filters Glass Fiber Filters
LabExact® Grade B Fine porosity, medium to fast flow rate, with a 1.0 µm size particle retention. Ideally suited for the collection of biochemical polymers that have been precipitated by denaturation. This material is twice as thick as Grade A with a higher loading capacity. May also be used as a solid particulate filter for gasses or as a prefilter.
  • Glass microfiber filters are produced from 100% borosilicate glass microfiber without the use of binders or strengthening agents
  • Depth filtration categorizes filter media that traps particulate within the
More Product Information