LabExact® Grade A-E 1.0 µm Binderless Glass Microfiber Filters
Supplier: I.W. TREMONT CO., INC.
LEAE4700 LEAE5500 LEAE7000 LEAE9000 LEAES810 LEAE1100 LEAE1250 LEAE1500 LEAE2100 LEAE2400 LEAE2500 LEAE3500 LEAE3700 LEAE4250
77590-406BX 37.12 USD
77590-406 77590-408 77590-410 77590-412 77590-414 77590-388 77590-390 77590-392 77590-394 77590-396 77590-398 77590-400 77590-402 77590-404
LabExact® Grade A-E 1.0 µm Binderless Glass Microfiber Filters
Filters Glass Fiber Filters
LabExact® Grade A-E fine porosity and fast flow rate, with a 1.0 µm size particle retention. DOP efficiency is 99.98%. Primarily used in suspended solids and air monitoring.
  • Glass microfiber filters are produced from 100% borosilicate glass microfiber without the use of binders or strengthening agents
  • Depth filtration categorizes filter media that traps particulate within the random matrix of fibers found within the thickness of the media
  • The efficiency rating of the filter is characteristic of the complex void space structure that develops as the fibers
More Product Information