IntelliStack™ Incubator Shakers, Single Unit
Supplier: Thermo Fisher Scientific
8882610D 8882610U 8885010D 8885010U
77590-030EA 23229 USD
77590-030 77590-032 77590-034 77590-036
IntelliStack™ Incubator Shakers, Single Unit
Shakers and Mixers Shaking Incubators
The Thermo Scientific™ IntelliStack™ Incubator shaker is a versatile laboratory instrument used for incubating and agitating samples simultaneously. It combines the functions of an incubator and shaker, providing a controlled environment for cell culture, microbial growth, and various other applications. With adjustable temperature, speed, and humidity settings, it offers precise control over experimental conditions. A large, easy-to-use 10" color touchscreen display with real-time monitoring of key parameters and alarms, alerts, and data logging with export via USB or Ethernet allows easyMore Product Information