Certified Screw Cap Mircotubes
Supplier: SARSTEDT INC
72.694.406 72.694.416 72.703.217 72.703.406 72.703.416 72.730.217 72.693.465 72.694.217 72.730.406 72.730.416 72.692.210 72.692.405 72.692.415
77588-824CS 455.31 USD
77588-824 77588-826 77588-882 77588-884 77588-886 77588-956 77588-790 77588-800 77588-976 77588-978 77588-774 77588-776 77588-778
Certified Screw Cap Mircotubes
Tubes Microtubes
SARSTEDT certfied screw cap microtubes provide the next level of security and quality for your every need.
  • Product quality tailored to specific applications ensures maximum reproducibility
  • Certified and guaranteed compliance values for PCR Performance Tested and Biosphere® plus certified products
  • Robust polypropylene material for safety and versatility
  • Leak-resistant according to ADR & IATA- tested at a vacuum of 0.95 bar
  • Centrifugation up to 20000×g
  • Can be autoclaved to 121 °C and frozen to -80 °C
  • O-ring cap increases cap security (IATA tested)
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable