SteriWare® LiquiDip Samplers
Supplier: SAMPLING SYSTEMS LTD
8921H-1100 8921H-1100S 8921H-450 8921H-450S
77577-556EA 271.04 USD
77577-556 77577-558 77577-560 77577-562
SteriWare® LiquiDip Samplers
Samplers Sample Dippers
The LiquiDip samplers are designed for taking cross sectional or point samples of liquids. The valve on the base of the sampler automatically closes when it is pushed against the container.
  • Cleanroom assembly and packing, ensuring cleanliness, in a class 100000 cleanroom
  • Can be used straight-from-the-bag: No risk of cross contamination
  • BSE-/TSE-free
LiquiDip Mono is suitable for taking cross sectional samples, by simply pushing to the bottom of the container.
LiquiDip Duo can be used as a transport container when both valves are closed for bothMore Product Information