SteriWare® V Sampling Spatulas with Product Retention
Supplier: SAMPLING SYSTEMS LTD
8115A-100 8115A-100S 8115A-150 8115A-150S 8115A-250 8115A-250S 8115N-100 8115N-100S 8115N-150 8115N-150S 8115N-250 8115N-250S
77577-074EA 170.15 USD
77577-074 77577-076 77577-078 77577-080 77577-082 77577-084 77577-086 77577-088 77577-090 77577-092 77577-094 77577-096
SteriWare® V Sampling Spatulas with Product Retention
Spatulas
These V spatulas are ideal for goods in and general powder sampling. The angled tip design keeps the powder in the sampler which is perfect for almost all powders. These spatulas can also be used for harder more viscous materials.
  • Robust PS construction
  • Available in 3 volumes - 100, 150 and 250 mm blade lengths
  • Individually bagged and produced in a cleanroom
  • Full batch traceability
2 widths are available. The 10 mm is a narrow design ideal for sampling small amounts from bottles. The 20 mm is the standard design for sampling from sacks, bags, kegs andMore Product Information