SteriWare® Slot Samplers
Supplier: SAMPLING SYSTEMS LTD
8073H-500 8073H-500S 8073H-1000 8073H-1000S 8073H-300 8073J-1000 8073J-1000S
77576-986EA 270.51 USD
77576-986 77576-988 77576-980 77576-982 77576-984 77576-990 77576-992
SteriWare® Slot Samplers
Samplers Disposable Samplers
These cross sectional multilevel disposable powder samplers are designed for powders. The robust HDPE construction make this to be used in huge number of testing applications.
  • BSE/TSE-free
  • Full batch traceability
  • Suitable for use in ATEX zones 0, 1 and 2
  • Available as gamma irradiated sterile (SAL 10⁻⁶)
Can take a composite from the top, middle and bottom of the powder bed - Moulded and packed in a cleanroom to ensure cleanliness.
Ideal for taking a multilevel, cross sectional sample from powders and granules. Insert the sampler and rotate theMore Product Information