Thermal Transfer Biopsy Cassettes, PiSmart
Supplier: StatLab
CAS-PI100BX-OR CAS-PI100BX-PK CAS-PI100BX-YL CAS-PI100BX-TN CAS-PI100BX-LV CAS-PI100BX-WH CAS-PI100BX-GR CAS-PI100BX-GY CAS-PI100BX-BL
77575-626CS 747.94 USD
77575-626 77575-628 77575-630 77575-632 77575-634 77575-618 77575-620 77575-622 77575-624
Thermal Transfer Biopsy Cassettes, PiSmart
Embedding Cassettes Biopsy Processing Cassettes
Two-piece design with a separate lid and square 0.9 mm pores.
  • Made from acetal polymer specifically validated for the PiSmart cassette printer
  • Compatible with the PiSmart Thermal Transfer Cassette Printer
  • Threaded
  • Nine colors
Pi Cassettes feature four raised pips in the corners of each print face to protect printed surface during dewaxing, while enabling the surface to be cleared of wax.
The new and improved cassettes now incorporate a smoother print face, resulting in improved print of barcodes and text. This improvement also means you can use aMore Product Information