CleanDox Cleanroom Papers
Supplier: Veltek Associates
C2WR-CD8.5X11-01 C2WR-CD8.5X11-BL-01 C2WR-CD8.5X11-CR-01 C2WR-CD8.5X11-GR-01 C2WR-CD8.5X11-PK-01
77575-340CS 269.7 USD
77575-340 77575-336 77575-338 77575-332 77575-334
CleanDox Cleanroom Papers
Paper
CleanDox is a poly-infused bond paper that can be used throughout your facility for note taking, printing batch records, forms and logbooks.
  • Non sterile
  • This IPA resistant material is available in several different colors
Packaging: 2500 sheets per case.