VWR® Multi-Channel Polypropylene Reagent Reservoirs
Supplier: VWR International
77575-288 77575-290 77575-292 77575-294 77575-296 77575-298 77575-300 77575-302
77575-288CS 73.24 USD
77575-288 77575-290 77575-292 77575-294 77575-296 77575-298 77575-300 77575-302
VWR® Multi-Channel Polypropylene Reagent Reservoirs
Reagent Reservoirs
These Polypropylene multi-channel reagent reservoirs are designed to support both automated liquid handling platforms and manual systems. The reservoir design complies with ANSI/SLAS 1-2004, when used for cell culture and immunoassay experiments.
  • Multiple capacities and well numbers can be selected for different experimental needs
  • Multi-channel reagent reservoirs are suitable for both 8-channel and 12-channel pipettes
  • Rhombic well series: 96- or 384-well reagent reservoirs on the bottom
  • Non-sterile or sterilized by irradiation, SAL 10⁻⁶ (ISO 11137)
  • Lot-
More Product Information