Controllers for Rotavapor® R-80 Rotary Evaporators, I-80/I-180
Supplier: Buchi
11I1801 11I1802 11I1803 11I0800 11I0801 11I0802 11I1800
77569-858EA 1969.14 USD
77569-858 77569-860 77569-862 77569-850 77569-852 77569-854 77569-856
Controllers for Rotavapor® R-80 Rotary Evaporators, I-80/I-180
Pumps Vacuum Controllers
The I-80 and I-180 are easy to use interfaces to conveniently regulate vacuum manually or automatically. Increase control of the evaporation process by setting the desired vacuum and accessing other modes of operation.
  • Functional design fulfils essential needs of vacuum regulation
  • Saves resources thanks to control of vacuum pump
  • High longevity due to robust and durable design
  • User-friendly and intuitive handling due to self-explaining user interface
  • Easy and accurate vacuum regulation
  • Relevant information is displayed at a glance
  • High reliability due to
More Product Information
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable