PureClean™ Tyvek® 1421B Filling Needle Autoclave Bags with Drawstring
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
6STK0101TD 6STK0102TD 6STK0103TD 6STK0105TD 6STK0106TD 6STK0107TD 6STK0108TD 6STK0109TD 6STK0111TD 6STK01-502TD 6STK0310TD 6STK0312TD 6STK0314TD 6STK0709TD 6STK406TWTD 6STK01-505TD 6STK0202TD 6STK0203TD 6STK0204TD 6STK0205TD 6STK0206TD 6STK0207TD 6STK0208TD 6STK0209TD 6STK0210TD 6STK0211TD 6STK0212TD 6STK0213TD 6STK02-503TD 6STK0304TD 6STK0305TD 6STK0306TD 6STK0308TD 6STK0309TD
77530-908CS 2095.5 USD
77530-908 77530-896 77530-940 77530-906 77530-912 77530-880 77530-900 77530-894 77530-926 77530-916 77530-892 77530-920 77530-930 77530-936 77530-884 77530-878 77530-886 77530-890 77530-918 77530-910 77530-874 77530-888 77530-902 77530-934 77530-944 77530-924 77530-932 77530-904 77530-942 77530-914 77530-882 77530-898 77530-938 77530-922
PureClean™ Tyvek® 1421B Filling Needle Autoclave Bags with Drawstring
Bags Sterilization Pouches
PureClean™ Autoclavable Filling Needle Bags are made from 1421B DuPont™ Tyvek®, which is a grade of uncoated, untreated, flexible Tyvek® material. It has been specifically designed for pharmaceutical use in steam sterilization applications. Keystone's PureClean™ Tyvek® material does not have a water soluble antistat agent applied to it during its manufacturing process.
  • Tear and puncture resistant
  • Eliminates need to cut material and the use of autoclave tape
  • Fast and consistent for equipment preparation area
  • Steam indicator
  • Hydrophobic
  • Lint-free
  • Strong
More Product Information