PureClean Tyvek® 1421B Self-Sealing Autoclave Bags with Steam Indicator
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
6STK0843SS 6STK09-512SS 6STK1015SS 6STK1018SS 6STK1020SS 6STK1022SS 6STK1024SS 6STK1030SS 6STK1036SS 6STK1048SS 6STK1056SS 6STK1062SS 6STK1212SS 6STK1214SS 6STK1216SS 6STK1218SS 6STK1220SS 6STK1222SS 6STK1224SS 6STK3238SS 6STK3248SS 6STK3412SS 6STK3434SS 6STK3456SS 6STK3624SS 6STK3628SS 6STK3636SS 6STK3642SS 6STK3646SS 6STK3648SS 6STK3660SS 6STK3664SS 6STK3672SS 6STK3838SS 6STK4024SS 6STK4040SS 6STK4048SS 6STK4054SS 6STK4060SS 6STK2044SS 6STK2046SS 6STK2048SS 6STK2134SS 6STK2224SS 6STK2226SS 6STK2240SS 6STK2260SS 6STK2424SS 6STK2430SS 6STK1604SS 6STK1612SS 6STK1616SS 6STK1620SS 6STK1624SS 6STK1628SS 6STK1630SS 6STK1632SS 6STK1635SS 6STK1636SS 6STK0608SS 6STK0610SS 6STK0612SS 6STK0615SS 6STK2834SS 6STK2840SS 6STK2844SS 6STK3016SS 6STK3020SS 6STK3028SS 6STK3030SS 6STK3036SS 6STK3060SS 6STK3232SS 6STK0638SS 6STK0707SS 6STK0709SS 6STK0713SS 6STK0811SS 6STK0815SS 6STK0816SS 6STK0820SS 6STK0838SS 6STK2436SS 6STK2440SS 6STK2448SS 6STK2452SS 6STK2462SS 6STK2628SS 6STK2634SS 6STK2646SS 6STK2828SS 6STK2832SS 6STK0404SS 6STK0406SS 6STK0409SS 6STK0410SS 6STK0416SS 6STK0426SS 6STK0428SS 6STK0430SS 6STK1418SS 6STK1420SS 6STK1424SS 6STK1426SS 6STK1436SS 6STK1438SS 6STK1440SS 6STK1448SS 6STK1526SS 6STK1543SS 6STK1230SS 6STK1232SS 6STK1234SS 6STK1236SS 6STK1240SS 6STK1242SS 6STK1246SS 6STK1248SS 6STK1414SS 6STK1416SS 6STK1848SS 6STK1854SS 6STK1870SS 6STK2020SS 6STK2024SS 6STK2026SS 6STK2030SS 6STK2032SS 6STK2036SS 6STK2040SS 6STK4072SS 6STK4242SS 6STK4244SS 6STK4416SS 6STK4450SS 6STK4848SS 6STK4850SS 6STK4864SS 6STK5054SS 6STK6250SS 6STK1228SS 6STK1638SS 6STK1644SS 6STK1648SS 6STK1820SS 6STK1824SS 6STK1828SS 6STK1830SS 6STK1832SS 6STK1836SS 6STK1840SS
77530-346CS 2763.75 USD
77530-346 77530-188 77530-394 77530-406 77530-158 77530-386 77530-226 77530-246 77530-186 77530-182 77530-340 77530-410 77530-152 77530-334 77530-128 77530-426 77530-156 77530-326 77530-116 77530-306 77530-194 77530-180 77530-132 77530-360 77530-304 77530-332 77530-414 77530-184 77530-336 77530-264 77530-140 77530-398 77530-266 77530-392 77530-278 77530-416 77530-126 77530-232 77530-290 77530-302 77530-170 77530-338 77530-376 77530-314 77530-166 77530-298 77530-366 77530-224 77530-358 77530-198 77530-368 77530-216 77530-370 77530-422 77530-252 77530-378 77530-320 77530-112 77530-420 77530-396 77530-228 77530-348 77530-234 77530-164 77530-110 77530-208 77530-124 77530-424 77530-250 77530-344 77530-138 77530-282 77530-408 77530-258 77530-150 77530-230 77530-318 77530-200 77530-196 77530-238 77530-202 77530-222 77530-296 77530-300 77530-374 77530-418 77530-330 77530-174 77530-388 77530-256 77530-404 77530-380 77530-276 77530-364 77530-120 77530-350 77530-262 77530-218 77530-286 77530-384 77530-146 77530-108 77530-382 77530-328 77530-354 77530-176 77530-148 77530-192 77530-212 77530-220 77530-190 77530-272 77530-280 77530-310 77530-242 77530-362 77530-260 77530-118 77530-248 77530-274 77530-352 77530-412 77530-142 77530-210 77530-178 77530-372 77530-308 77530-162 77530-136 77530-294 77530-236 77530-268 77530-144 77530-172 77530-342 77530-244 77530-324 77530-400 77530-206 77530-168 77530-284 77530-154 77530-122 77530-160 77530-402 77530-114 77530-288 77530-390 77530-214 77530-240 77530-204
PureClean Tyvek® 1421B Self-Sealing Autoclave Bags with Steam Indicator
Bags Sterilization Pouches
Self-sealing Tyvek® 1421B autoclave bags with steam indicators can be used as either the inner, or outer bag for the steam sterilization of pharmaceutical change parts.Self-sealing Tyvek® 1421B autoclave bags with steam indicators can be used as either the inner, or outer bag for the steam sterilization of pharmaceutical change parts.
  • Uncoated Tyvek® on both sides for maximum breathability during sterilization without the chance of residue deposits on equipment post sterilization
  • Tear and puncture resistant
  • Convenient and reliable Self-Seal Closure
More Product Information