Durx® 670 AS (Aerospace) Non-woven Cleanroom Wiper, 55% Cellulose/45% Polyester
Supplier: Berkshire
DR670090920AS DR670121220AS
77528-884CS 479.17 USD
77528-884 77528-876
Durx® 670 AS (Aerospace) Non-woven Cleanroom Wiper, 55% Cellulose/45% Polyester
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Durx® 670 AS for Aerospace (AS) is a hydroentangled, non-woven blend of 55% cellulose and 45% polyester with no binders or other chemical additives. This wiper meets the AMS 3819D, Class 2, Grade A, Form 1 requirements.
  • Excellent combination of the synthetic polyester strength and cleanliness, with the absorbent characteristics of cellulose
  • Smooth, highly absorbent, and durable
  • Low extractable levels and particle counts
  • Chemically compatible with common cleaning solutions
  • Economical
Durx® 670 AS for Aerospace (AS) is a hydroentangled nonwoven blend ofMore Product Information