Urea ACS, Ultrapure

Supplier: RPI

Synonyms: Carbonyl diamide, Carbamide, Diaminomethanone, Carbonyldiamide

U20225-3000.0 U20225-500.0 U20225-25000.0
76348-792EA 94.9 USD
76348-792 76348-794 77510-504
Urea ACS, Ultrapure
Urea

Caution: Research or further manufacturing use only, not for food or drug use.

Formula: NH₂CONH₂
MW: 60.06 g/mol
Boiling Pt: 197
Melting Pt: 132.7
Storage Temperature: Room Temperature
MDL Number: MFCD00008022
CAS Number: 57-13-6
EINECS: 200-315-5
Merck Index: 13,09935

Order Now

Specification Test Results

Appearance White Powder or Pellets
Assay (Titration, dried basis) 99.0 - 100.5%
Chlorides ≤5 ppm
Heavy Metals (as Pb) ≤0.001%
Insoluble Matter ≤0.01%
Melting Point 132 - 135 °C
Residue on Ignition ≤0.01%
Sulfate (SO4) Conforms
BSE/TSE Free BSE/TSE Free
Grade ACS


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR