Sodium acetate ACS

Supplier: RPI

Synonyms: Acetic acid sodium salt

S22045-500.0 S22045-10000.0
76348-086EA 31.52 USD
76348-086 77510-422
Sodium acetate ACS
Sodium acetate

Caution: Research or further manufacturing use only, not for food or drug use.

Formula: H₃CCOONa
Melting Pt: >300 °C
Storage Temperature: Room Temperature
MDL Number: MFCD00012459
CAS Number: 127-09-3
EINECS: 204-823-8

Order Now

Specification Test Results

Appearance White Powder
Assay (Titration) ≥ 99.0 %
Calcium (Ca) ≤ 0.005 %
Chloride Content ≤ 0.002 %
Heavy Metals ≤ 0.001 %
Infrared Spectrum Conforms to structure
Insoluble Matter ≤ 0.01 %
Iron (Fe) ≤ 0.001 %
Loss on Drying ≤ 1.0 %
Magnesium (Mg) ≤ 0.002 %
pH (5%, water) @25 °C 7.0 - 9.2
Phosphate ≤ 0.001 %
Grade ACS


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR